Läs mer
Tocris Bioscience™ RKI 1447 dihydrochloride
Potent and selective ROCK inhibitor; antitumor
Brand: Tocris Bioscience™ 5061/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
RKI 1447 dihydrochloride is a potent and selective ROCK inhibitor (IC50 values are 6.2 and 14.5 nM for ROCK2 and ROCK1 respectively); suppresses phosphorylation of ROCK substrates MLC-2 and MYPT-1 in human cancer cells, but has no effect on the phosphorylation levels of Akt, MEK, and S6 kinase. Displays anti-invasive and antitumor activities in breast cancer cells.Specifikationer
| >99% | |
| 1782109-09-4 | |
| 50 mg |
Produkttitel
For Research Use Only