missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMND5A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RMND5A |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16837474
|
Proteintech
17559-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16827474
|
Proteintech
17559-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RMND5A is a protein coding gene.Specifications
RMND5A | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q9H871 | |
64795 | |
RMND5A Fusion Protein Ag11751 | |
Primary | |
-20°C | |
RMND5A |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CTLH, p44CTLH, Protein RMD5 homolog A, RMD5, RMND5A | |
RMND5A | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |