Läs mer
RMP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Produktkod. 16848243 Handla allt Proteintech Produkter
Change view
Kvantitet:
20 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16848243 20 μL
16838243 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16848243 Leverantör Proteintech Leverantörsnummer 112771AP20UL
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Rabbit Polyclonal Antibody
The function of the C19orf2 protein remains unknown.The protein encoded by this gene binds to RNA polymerase II subunit 5 (RPB5) and negatively modulates transcription through its binding to RPB5. The encoded protein seems to have inhibitory effects on various types of activated transcription, but it requires the RPB5-binding region. This protein acts as a corepressor. It is suggested that it may require signaling processes for its function or that it negatively modulates genes in the chromatin structure. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifikationer
Antigen RMP
Användningsområden Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
Klassificering Polyclonal
Koncentration 0.2 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen URI1
Genaccessionsnr. O94763, Q3TLD5
Gene Alias C19orf2, NNX3, PPP1R19, Protein NNX3, RMP, RPB5 mediating protein, URI
Gensymboler URI1
Värd art Rabbit
Immunogen RMP Fusion Protein Ag1789
Reningsmetod Antigen Affinity Chromatography
Kvantitet 20 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 19777, 8725
Målarter Mouse, Human
Innehåll och lagring -20°C
Produkttyp Antibody
Form Liquid
Isotyp IgG