Läs mer
RN 1734, TRC
Produktkod. 30420210
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30420210 100 mg
30413360 250 mg
30419918 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30420210 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR637005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 946387-07-1
Molekylformel C14H22Cl2N2O2S
Molekylvikt (g/mol) 353.31
Synonym RN 1734, 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide
IUPAC-namn 2,4-dichloro-N-propan-2-yl-N-[2-(propan-2-ylamino)ethyl]benzenesulfonamide
LEDER CC(C)NCCN(C(C)C)S(=O)(=O)c1ccc(Cl)cc1Cl
Specifikationer
Kemiskt namn eller material 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide
Kvantitet 100 mg
InChI formel InChI=1S/C14H22Cl2N2O2S/c1-10(2)17-7-8-18(11(3)4)21(19,20)14-6-5-12(15)9-13(14)16/h5-6,9-11,17H,7-8H2,1-4H3
Formel vikt 352.0779