missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RNASEH1 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16834854
|
Proteintech
15606-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16824854
|
Proteintech
15606-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
RNASEH1 belongs to the RNase H family. It contains 1 RNase H domain. RNASEH1 is an endonuclease that specifically degrades the RNA of RNA-DNA hybrids.Specifications
RNASEH1 | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O60930, O70338, Q5BK46 | |
19819, 246243, 298933 | |
RNASEH1 Fusion Protein Ag7990 | |
Primary | |
-20°C | |
RNASEH1 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
H1RNA, Ribonuclease H type II, ribonuclease H1, RNase H1, RNASEH1, RNH1 | |
Rnaseh1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |