missing translation for 'onlineSavingsMsg'
Learn More

RND3 Mouse anti-Human, Porcine, Clone: 3H8F9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66228-1-IG-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16827915

  • 1236.20 SEK / 20µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras, Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex; GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins; and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form.
Specifications

Specifications

RND3
3H8F9
Unconjugated
RND3
Liquid
RND3
RND3 Fusion Protein Ag18839
Protein A
RUO
Antibody
Monoclonal
Porcine, Human
Western Blot
1.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O77683, P61587
ARHE, memB, Protein MemB, Rho family GTPase 3, Rho8, RhoE, RND3
Mouse
IgG2b
20 μL
-20°C
Primary
390, 397559
Videos
SDS
Documents
Certificates
Special Offers

Special Offers