missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RNF113A Recombinant Protein

Brand:  Abnova™ H00007737-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16122082

  • 6845.00 SEK / 25µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH00832
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
63.47
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAEQLSPGKAVDQVCTFLFKKPGRKGAAGRRKRPACDPEPGESGSSSDEGCTVVRPEKKRVTHNPMIQKTRDSGKQKAAYGDLSSEEEEENEPESLGVVYKSTRSAKPVGPEDMGATAVYELDTEKERDAQAIFERSQKIQEELRGKEDDKIYRGINNYQKYMKPKDTSMGNASSGMVRKGPIRAPEHLRATVRWDYQPDICKDYKETGFCGFGDSCKFLHDRSDYKHGWQIERELDEGRYGVYEDENYEVGSDDEEIPFKCFICRQSFQNPVVTKCRHYFCESCALQHFRTTPRCYVCDQQTNGVFNPAKELIAKLEKHRATGEGGASDLPEDPDEDAIPIT
RNF113/ZNF183
RNF113A
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
7737
RNF113A (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RNF113A
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers