missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ RNF3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA131805
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: The peptide used to generate this antibody is available for purchase (GTX23892-PEP).. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Phosphorylase kinase is a hexadecameric enzyme that is comprised of four copies of four subunits that are encoded by four separate genes: PHKA, PHKB, PHKG, and PH kDa. This serine/threonine specific kinase converts glycogen phosphorylase b to glycogen phosphorylase a, resulting in the release of glucose-1-phophate from glycogen. PHKB (Phosphorylase b kinase regulatory subunit beta) is a 1093 amino acid subunit of phosphorylase kinase that, along with PHKA, has regulatory functions controlled by phosphorylation. Defects in the gene encoding PHKB are the cause of glycogen storage disease type 9B, which is also known as phosphorylase kinase deficiency of liver and muscle. This disease is characterized by a mild phenotype of hepatomegaly with only slightly elevated transaminase and plasma lipids, no clinical muscle involvement, and generally is correlated with a gradual improvement with increasing age. There are four isoforms of PHKB that are produced as a result of alternative splicing events.
Specifikationer
| RNF3 | |
| Polyclonal | |
| Unconjugated | |
| PCGF3 | |
| 2310035N15Rik; AI662857; D630042K08Rik; D6S113E; DADB-17J1.3; DKFZp686D20235; DKFZp686N0336; DONG1; E430039C14; FLJ31942; FLJ36550; FLJ43813; FSH; FSRG1; KIAA9001; MGC129075 protein; MGC129615; MGC40413; NAT; PCGF3; polycomb group ring finger 3; polycomb group RING finger protein 3; ring finger protein 3; RING finger protein 3A; RING3; RNF3; RNF3A | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 10336 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q3KNV8 | |
| PCGF3 | |
| Peptide with sequence C-PLLLHYRPKMDLL, from the C Terminus. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering