Läs mer
RNF43 Antibody (923227R), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7964J
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
RNF43 Monoclonal antibody specifically detects RNF43 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifikationer
| RNF43 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 923227R | |
| Flow Cytometry, CyTOF-ready | |
| DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC | |
| Human embryonic kidney cell line HEK293-derived recombinant human RNF43, Glu43-Val199, Accession # Q68DV7 | |
| 0.1 mL | |
| Cancer | |
| 54894 | |
| Store at 4°C in the dark. | |
| IgG1 |