Läs mer
Ro 32-3555, TRC
Produktkod. 30424366
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424366 10 mg
30410952 2.5 mg
30422276 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424366 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70090510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 190648-49-8
Molekylformel C22H36N4O5
Molekylvikt (g/mol) 436.55
Synonym (alphaR,betaR)-beta-(Cyclopentylmethyl)-N-hydroxy-gamma-oxo-alpha-[(3,4,4-trimethyl-2,5-dioxo-1-imidazolidinyl)methyl]-1-piperidinebutanamide, Cipemastat, Ro 32-3555/000, Trocade, Ro 32-3555
IUPAC-namn (2R,3R)-3-(cyclopentylmethyl)-N-hydroxy-4-oxo-4-piperidin-1-yl-2-[(3,4,4-trimethyl-2,5-dioxoimidazolidin-1-yl)methyl]butanamide
LEDER CN1C(=O)N(C[C@@H]([C@@H](CC2CCCC2)C(=O)N3CCCCC3)C(=O)NO)C(=O)C1(C)C
Specifikationer
Kemiskt namn eller material [R-(R*,R*)]-beta-(Cyclopentylmethyl)-N-hydroxy-gamma-oxo-alpha-[(3,4,4-trimethyl-2,5-dioxo-1-imidazolidinyl)methyl]-1-piperidinebutanamide
Kvantitet 10 mg
InChI formel InChI=1S/C22H36N4O5/c1-22(2)20(29)26(21(30)24(22)3)14-17(18(27)23-31)16(13-15-9-5-6-10-15)19(28)25-11-7-4-8-12-25/h15-17,31H,4-14H2,1-3H3,(H,23,27)/t16-,17+/m1/s1
Formel vikt 436.2686