Läs mer
Tocris Bioscience™ Ro 48-8071 fumarate
GREENER_CHOICE
2,3-Oxidosqualene cyclase (OSC) inhibitor; blocks cholesterol synthesis
Brand:  Tocris Bioscience™ 5389/50
Encompass
Produktkod. 15746786
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
Ro 48-8071 fumarate is a 2,3-Oxidosqualene cyclase (OSC) inhibitor (IC50 = 6.5 nM); blocks cholesterol synthesis in HepG2 cells. Reduces viability of breast cancer cell lines. Also inhibits Ebola virus (EBOV) cell entry (IC50 = 1.74 μM).
Specifikationer
Specifikationer
99%
189197-69-1
50 mg
(4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone
4°C
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer
Produkttitel
Välj ett problem
For Research Use Only