Läs mer
Tocris Bioscience™ Ro 48-8071 fumarate
Beskrivning
Ro 48-8071 fumarate is a 2,3-Oxidosqualene cyclase (OSC) inhibitor (IC50 = 6.5 nM); blocks cholesterol synthesis in HepG2 cells. Reduces viability of breast cancer cell lines. Also inhibits Ebola virus (EBOV) cell entry (IC50 = 1.74 μM).
Specifikationer
Specifikationer
| CAS | 189197-69-1 |
| Kvantitet | 50 mg |
| Molekylformel | (4-Bromophenyl)[2-fluoro-4-[[6-(methyl-2-propen-1-ylamino)hexyl]oxy]phenyl]methanone |
| Produkttyp | Ro 48-8071 fumarate |
| Renhet | 99% |
| Molekylvikt (g/mol) | M.W. (Observed): 564.44 |
| Inhibitorer | Ro 48-8071 fumarate |
For Research Use Only