Läs mer
Tocris Bioscience™ Ro 64-5229
GREENER_CHOICE
Produktkod. 18044382
Change view
Click to view available options
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
18044382 50 mg
18024602 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18044382 Supplier Tocris Bioscience™ Supplier No. 2913/50
Selective, non-competitive mGlu2 antagonist
Selective, non-competitive mGlu2 antagonist. Inhibits GTPγ35S binding to mGlu2-containing membranes (IC50 = 0.11 μM).
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 246852-46-0
Molekylformel C17H19Cl2N3O
Specifications
Kemiskt namn eller material Ro 64-5229
Kvantitet 50 mg
Formel vikt 352.26
Renhet >99%
Molekylvikt (g/mol) 352.259