Learn More
RO-9187, MedChemExpress
Product Code. 16456947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16456947 5 mg 5mg
16444228 10 mg 10mg
16440567 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16456947 Supplier Medchem Express Supplier No. HY10870
 Restricted Item
Beräknad leverans: 23-11-2026
MedChemExpress RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.
RO-9187 is a potent inhibitor of HCV virus replication with an IC50 of 171 nM.
Specifications
Chemical Name or Material RO-9187
CAS 876708-03-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C9H12N6O5
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (351.83 mM; Need ultrasonic) ∣H2O : 7.14 mg/mL (25.12 mM; Need ultrasonic)
SMILES O=C1N=C(N)C=CN1[C@H]2[C@@H](O)[C@H](O)[C@](CO)(N=[N+]=[N-])O2
Molecular Weight (g/mol) 284.23
Formula Weight 284.23
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus