Läs mer
RO4929097, MedChemExpress
Produktkod. 16485286
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16485286 100 mg -
16447117 5 mg 5mg
16424398 10 mg -
16420717 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16485286 Leverantör Medchem Express Leverantörsnummer HY11102
MedChemExpress RO4929097 (RG-4733) is a γ secretase inhibitor with IC50 of 4 nM, inhibiting cellular processing of Aβ40 and Notch with EC50 of 14 nM and 5 nM, respectively.
RO4929097 (RG-4733) is a γ secretase inhibitor with IC50 of 4 nM, inhibiting cellular processing of Aβ40 and Notch with EC50 of 14 nM and 5 nM, respectively.
Specifikationer
Kemiskt namn eller material RO4929097
Fysisk form Solid
CAS 847925-91-1
Färg White
Kvantitet 100 mg
Anteckningar om renhetsgrad Research
Molekylformel C22H20F5N3O3
För användning med (applikation) Cancer-programmed cell death
Synonym RG-4733
Löslighetsinformation DMSO : ≥ 49 mg/mL (104.39 mM)
LEDER O=C(C(C)(C(NCC(F)(C(F)(F)F)F)=O)C)N[C@H]1C2=CC=CC=C2C3=CC=CC=C3NC1=O
Molekylvikt (g/mol) 469.4
Formel vikt 469.4
Procent renhet 97.93%