Läs mer
Robenacoxib, TRC
Produktkod. 17972525
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17972525 50 mg
17954065 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17972525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR63800050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 220991-32-2
Molekylformel C16 H13 F4 N O2
Molekylvikt (g/mol) 327.27
Synonym Robenacoxib, 5-Ethyl-2-[(2,3,5,6-tetrafluorophenyl)amino]benzeneacetic Acid;
IUPAC-namn 2-[5-ethyl-2-(2,3,5,6-tetrafluoroanilino)phenyl]acetic acid
LEDER CCc1ccc(Nc2c(F)c(F)cc(F)c2F)c(CC(=O)O)c1
Specifikationer
Kemiskt namn eller material Robenacoxib
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C16H13F4NO2/c1-2-8-3-4-12(9(5-8)6-13(22)23)21-16-14(19)10(17)7-11(18)15(16)20/h3-5,7,21H,2,6H2,1H3,(H,22,23)
Formel vikt 327.0882
Procent renhet >95%