Learn More
Abnova™ ROBO1 Pre-design Chimera RNAi
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants (NM_001145845.1,NM_002941.3,NM_133631.3)
Brand: Abnova™ H00006091-R03.20nmol
Additional Details : Weight : 0.00010kg
Description
- Gene name: ROBO1
- Gene Alias: DUTT1, FLJ21882, MGC131599, MGC133277, SAX3
- Entrez GeneID: 6091
- strong>Gene Description: Roundabout, axon guidance receptor, homolog 1
Specifications
RNAi Knockdown | |
Homo sapiens roundabout, axon guidance receptor, homolog 1 (Drosophila) (ROBO1), transcript variants, mRNA. This product targets for all of 3 transcript variants. (NM_001145845.1,NM_002941.3,NM_133631.3) | |
ROBO1 Pre-design Chimera RNAi | |
Store at -20°C, do not exceed 4 - 5 freeze-thaw cycles to ensure product integrity. |
Human | |
DEPC water | |
20 nmol | |
Human |
For research use only