missing translation for 'onlineSavingsMsg'
Learn More

ROBO1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25181-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16823745

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specialized cells at the midline, which separates the left and right halves of the CNS, have a number of roles in directing growth cone behavior. In the vertebrate spinal cord, the insect ventral nerve cord and in C. elegans, midline cells produce guidance cues such as nectins and slit, which act as attractants and repellents, respectively. These cells may act as gatekeepers to prevent axons from crossing the midline and to induce a switch in growth cone responsiveness to guidance cues beyond the gateway. One such gatekeeper, Robo, is an axon guidance receptor that defines a novel subfamily of Ig superfamily proteins that are conserved from fruit flies to mammals. Robo acts as a receptor for the repellent Slit and functions in a cell-autonomous fashion. Non-crossing axons express high levels of Robo, whereas crossing axons express low levels of Robo before reaching the midline and high levels after they cross. Robo1 and Robo2 are two human homologs of the Drosophila protein Roundabout. Robo1 is also homologous to the C. elegans gene sax3, whereas Robo2 is homologous to the zebrafish gene astray.
Specifications

Specifications

ROBO1
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y6N7
Deleted in U twenty twenty, DUTT1, H Robo 1, ROBO1, Roundabout homolog 1, SAX3
Rabbit
IgG
20 μL
-20°C
Primary
6091
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ROBO1
Liquid
ROBO1
ROBO1 Fusion Protein Ag16461
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers