missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ROCK2, GST Tag Recombinant Protein
ROCK2 Recombinant Human Protein
5380.00 SEK
Specifications
Accession Number | NP_004841 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 90.8 kDa |
Gene Alias | ROCK2 |
KinaseFamily | DMPK Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11573517
|
Invitrogen
PV3759 |
10μg |
5380.00 SEK
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ROCK2, the predominant ROCK isoform in skeletal muscle, is an important regulator downstream of Rho GTPases.Specifications
NP_004841 | |
90.8 kDa | |
DMPK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
Kinase Assay | |
ROCK2 | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: ACG Group | |
Baculovirus | |
Recombinant, Catalytic |