missing translation for 'onlineSavingsMsg'
Learn More

ROGDI Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17047-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16876864

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a protein of unknown function. Loss-of-function mutation in this gene cause Kohlschutter-Tonz syndrome. Alternate splicing results in multiple transcript variants.
Specifications

Specifications

ROGDI
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3TDK6, Q9GZN7
Protein rogdi homolog, ROGDI, rogdi homolog (Drosophila)
Rabbit
IgG
20 μL
-20°C
Primary
66049, 79641
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
Unconjugated
ROGDI
Liquid
Rogdi
ROGDI Fusion Protein Ag10688
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers