Learn More
Heidolph™ Rollers
Product Code. 10613031 Shop All Heidolph Instruments Products
Change view
Click to view available options
Length (Metric):
258 mm
Unit Size:
Each
Product Code. Length (Metric) Unit Size
10613031 258 mm Each
1 options
Product Code. 10613031 Supplier Heidolph™ Supplier No. 5497000000
  • ONLINE EXCLUSIVE PRICE
    3435.00 SEK
    3204.84 SEK / Paket med 1
    Save 230.16 SEK
    6% Av
Beräknad leverans: 01-12-2026
Heidolph frame with 2 tension rollers ideally used in research, scientific laboratory and other environments.
  • Spare tension roller
  • 1 tension roller as accessory for attachment 549-70000-00
  • Accessory category D for Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
Order Info
Size
TRUSTED_SUSTAINABILITY
Specifications
Voltage 115/230 V
Product Type Rollers
Length (Metric) 258 mm