missing translation for 'onlineSavingsMsg'
Läs mer

Ropinirole-d7 Hydrochloride, TRC

Produktkod. 30419013
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30419013 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30419013 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCR6410041MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Ropinirole hydrochloride N-propyl-d7
CAS 1261396-31-9
Molekylformel C16 2H7 H17 N2 O . Cl H
Kvantitet 1 mg
Synonym Ropinirole-D7 hydrochloride, Ropinirole-D7 hydrochloride (N-propyl D7), 4-[2-[1,1,2,2,3,3,3-Heptadeuteriopropyl(propyl)amino]ethyl]-1,3-dihydro-2H-indol-2-one hydrochloride
InChI formel InChI=1S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H/i1D3,3D2,9D2;
LEDER Cl.[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])N(CCC)CCc1cccc2NC(=O)Cc12
IUPAC-namn 4-[2-[1,1,2,2,3,3,3-heptadeuteriopropyl(propyl)amino]ethyl]-1,3-dihydroindol-2-one;hydrochloride
Molekylvikt (g/mol) 303.88
Formel vikt 303.2095
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.