Läs mer
Ropinirole Hydrochloride, TRC
Produktkod. 30132749
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132749 100 mg
30132662 25 mg
17954315 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30132749 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR641000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91374-20-8
Molekylformel C16 H24 N2 O . Cl H
Molekylvikt (g/mol) 296.84
Synonym 2H-Indol-2-one, 4-[2-(dipropylamino)ethyl]-1,3-dihydro-, hydrochloride (1:1), 2H-Indol-2-one, 4-[2-(dipropylamino)ethyl]-1,3-dihydro-, monohydrochloride (9CI), Adartrel, Parkirop, Ralnea XL, Repreve, Requip, Ronirol, Ropark, Ropinirole hydrochloride, Ropitar, SKF 101468 hydrochloride, SKF 101468A, Spiroco XL
IUPAC-namn 4-[2-(dipropylamino)ethyl]-1,3-dihydroindol-2-one;hydrochloride
LEDER Cl.CCCN(CCC)CCc1cccc2NC(=O)Cc12
Specifikationer
Kemiskt namn eller material Ropinirole Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C16H24N2O.ClH/c1-3-9-18(10-4-2)11-8-13-6-5-7-15-14(13)12-16(19)17-15;/h5-7H,3-4,8-12H2,1-2H3,(H,17,19);1H
Formel vikt 296.1655