Läs mer
Roquefortine C, TRC
Produktkod. 30138664
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138664 5 mg -
17921515 2.5 mg 2.5mg
17923795 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138664 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR7008755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58735-64-1
Molekylformel C22 H23 N5 O2
Molekylvikt (g/mol) 389.45
Synonym 2H-Pyrazino[1',2':1,5]pyrrolo[2,3-b]indole-1,4(3H,5aH)-dione, 10b-(1,1-dimethyl-2-propenyl)-6,10b,11,11a-tetrahydro-3-(1H-imidazol-4-ylmethylene)-, (3E,5aS,10bR,11aS)- (9CI), (3E,5aS,10bR,11aS)-10b-(1,1-Dimethyl-2-propen-1-yl)-6,10b,11,11a-tetrahydro-3-(1H-imidazol-5-ylmethylene)-2H-pyrazino[1',2':1,5]pyrrolo[2,3-b]indole-1,4(3H,5aH)-dione, (-)-Roquefortine C, NSC 292134, Roquefortin, Roquefortine, Roquefortine C
LEDER CC(C)(C=C)[C@@]12C[C@@H]3N([C@@H]1Nc4ccccc24)C(=O)\C(=C/c5cnc[nH]5)\NC3=O
Specifikationer
Kemiskt namn eller material Roquefortine C
Kvantitet 5 mg
InChI formel InChI=1S/C22H23N5O2/c1-4-21(2,3)22-10-17-18(28)25-16(9-13-11-23-12-24-13)19(29)27(17)20(22)26-15-8-6-5-7-14(15)22/h4-9,11-12,17,20,26H,1,10H2,2-3H3,(H,23,24)(H,25,28)/b16-9+/t17-,20-,22+/m0/s1
Formel vikt 389.185