missing translation for 'onlineSavingsMsg'
Learn More

ROR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20629-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840195

  • 3313.85 SEK / 150µL
Estimated Shipment: 03-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ROR1 is a receptor protein tyrosine kinase whose cellular role has not been determined. It is a type I membrane protein and belongs to the ROR subfamily of cell surface receptors. Studies of a similar protein in mouse suggest that this protein may interact with another receptor protein tyrosine kinase and may be involved in skeletal and cardiac development.
Specifications

Specifications

ROR1
Polyclonal
Unconjugated
ROR1
NTRKR1, ROR1
Rabbit
Antigen Affinity Chromatography
RUO
26563, 362550, 4919
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q01973, Q9Z139
Ror1
ROR1 Fusion Protein Ag14586
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers