missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROS1 Recombinant Human Protein
ROS1 Recombinant Human Protein
Brand: Invitrogen PV3814
Additional Details : Weight : 0.05250kg
Description
ROS1 is a tyrosine kinase that functions as a growth or differentiation factor receptor.Specifications
NP_002935 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
79.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
ROS1, ROS, MCF4 | |
ROS1 was pre-diluted in enzyme dilution buffer and assayed in 25mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 670μg/mL Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Amino acids 1883-2347 | |
4806nmoles of phosphate transferred to Abl1 peptide substrate (EAIYAAPFAKKK) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 0.104μg/mL | |
GST Tag | |
Kinase and inhibitors | |
80% by SDS-PAGE |
0.36mg/mL | |
6098 | |
ROS1 Recombinant Human Protein | |
–80°C | |
1 Each | |
ROS1 Recombinant Human Protein | |
SEV | |
ROS1 Recombinant Human Protein | |
Human | |
Baculovirus | |
Receptor tyrosine kinases |
For Research Use Only. Not for use in diagnostic procedures.