Läs mer
STEMCELL Technologies RosetteSep™ Human CD3 Depletion Cocktail
Produktkod. 17107701
Encompass
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
200 mL whole blood
40 mL whole blood
Förpackningsstorlek:
1 uppsättning
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17107701 40 mL whole blood -
17157701 200 mL whole blood 1 uppsättning
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17107701 Leverantör STEMCELL Technologies Leverantörsnummer 15621
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Immunodensity depletion of CD3+ T cells
  • The RosetteSep™ Human CD3 Depletion Cocktail is designed to deplete CD3+ cells from whole blood
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD3 and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The CD3-depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium
Specifikationer
Celltyp T Cells
För användning med (applikation) Cell Isolation
Kvantitet 40 mL whole blood