Läs mer
Rosmarinic Acid, TRC
Produktkod. 17947834
Change view
Kvantitet:
10 mg
50 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17947834 10 mg
17948694 50 mg
17989454 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17947834 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR68480010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20283-92-5
Molekylformel C18 H16 O8
Molekylvikt (g/mol) 360.31
Synonym Rosmarinic Acid, Benzenepropanoic acid, α-[[(2E)-3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-, (αR)- (9CI), Benzenepropanoic acid, α-[[3-(3,4-dihydroxyphenyl)-1-oxo-2-propenyl]oxy]-3,4-dihydroxy-, [R-(E)]-, Cinnamic acid, 3,4-dihydroxy-, 2-ester with 3-(3,4-dihydroxyphenyl)lactic acid, Rosmarinic acid, Labiatenic acid, Labiatic acid, Mamorekku RUH 2, Meiji Red Perilla Polyphenol, NPLC 0542, RM 21A, Rosemaric acid, Rosemary acid, trans-Rosmarinic acid
IUPAC-namn (2R)-3-(3,4-dihydroxyphenyl)-2-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxypropanoic acid
LEDER OC(=O)[C@@H](Cc1ccc(O)c(O)c1)OC(=O)\C=C\c2ccc(O)c(O)c2
Specifikationer
Kemiskt namn eller material Rosmarinic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C18H16O8/c19-12-4-1-10(7-14(12)21)3-6-17(23)26-16(18(24)25)9-11-2-5-13(20)15(22)8-11/h1-8,16,19-22H,9H2,(H,24,25)/b6-3+/t16-/m1/s1
Formel vikt 360.0845
Procent renhet >95%