missing translation for 'onlineSavingsMsg'
Learn More

Rosuvastatin, calcium salt, 98%, ACROS Organics™

Brand:  Acros Organics 456230010

Additional Details : CAS Number : 147098-20-2 Weight : 0.00100kg


Quantity 1g

 View more versions of this product

Product Code. 15575916

  • 3175.00 SEK / 1g


Estimated Shipment
09-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

147098-20-2
500.58
3s,5r-rosuvastatin calcium salt, ent-rosuvastatin
calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid;molecular hydrogen
C22H28FN3O61/2Ca
LQFSLFAVBQEFBH-LSMDDVBQSA-N
131673834
[HH].[HH].CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Ca]

Specifications

Rosuvastatin, calcium salt
Glass Bottle
147098-20-2
C22H28FN3O61/2Ca
LQFSLFAVBQEFBH-LSMDDVBQSA-N
calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid;molecular hydrogen
131673834
98%
Authentic
1g
97.5% min. (HPLC)
3s,5r-rosuvastatin calcium salt, ent-rosuvastatin
[HH].[HH].CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.CC(C)C1=NC(=NC(=C1C=CC(CC(CC(=O)O)O)O)C2=CC=C(C=C2)F)N(C)S(=O)(=O)C.[Ca]
500.58
500.58
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Rosuvastatin calcium salt

Signal Word
  • Warning

Hazard Category
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 2

Hazard Statement
  • H362-May cause harm to breast-fed children.
  • H361d-Suspected of damaging the unborn child.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P263-Avoid contact during pregnancy/while nursing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents