Läs mer
Rosuvastatin Calcium Salt, TRC
Produktkod. 18090300
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18090300 10 mg
18050600 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18090300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70050010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 147098-20-2
Molekylformel 2 C22 H27 F N3 O6 S . Ca
Molekylvikt (g/mol) 1001.14
Synonym 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, calcium salt (2:1), (3R,5S,6E)-, 6-Heptenoic acid, 7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-, calcium salt (2:1), [S-[R*,S*-(E)]]-, Crestomed, Crestor, Fortius, Rostar, Rosuvastatin calcium, Rosuvastatin calcium salt, Rosuvastatin hemicalcium, Rozavel, S 4522, Suvikan, ZD 4522, calcium salt
IUPAC-namn calcium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
LEDER [Ca+2].CC(C)c1nc(nc(c1\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c2ccc(F)cc2)N(C)S(=O)(=O)C.CC(C)c3nc(nc(c3\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c4ccc(F)cc4)N(C)S(=O)(=O)C
Specifikationer
Kemiskt namn eller material Rosuvastatin Calcium
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/2C22H28FN3O6S.Ca/c2*1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h2*5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;;+2/p-2/b2*10-9+;/t2*16-,17-;/m11./s1
Formel vikt 1000.2835
Procent renhet >95%