Läs mer
Rosuvastatin Ethyl Ester, TRC
Produktkod. 30007146
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007146 1 g
17953875 100 mg
17982265 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007146 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR7007231G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 851443-04-4
Molekylformel C24 H32 F N3 O6 S
Molekylvikt (g/mol) 509.59
Synonym (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-6-heptenoic Acid Ethyl Ester, Rosuvastatin Ethyl Ester
IUPAC-namn ethyl (E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
LEDER CCOC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc(nc1c2ccc(F)cc2)N(C)S(=O)(=O)C)C(C)C
Specifikationer
Kemiskt namn eller material Rosuvastatin Ethyl Ester
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C24H32FN3O6S/c1-6-34-21(31)14-19(30)13-18(29)11-12-20-22(15(2)3)26-24(28(4)35(5,32)33)27-23(20)16-7-9-17(25)10-8-16/h7-12,15,18-19,29-30H,6,13-14H2,1-5H3/b12-11+/t18-,19-/m1/s1
Formel vikt 509.1996
Procent renhet >95%