Läs mer
Rosuvastatin Lactone, TRC
Produktkod. 17932135
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17932135 100 mg
17954405 10 mg
17920905 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17932135 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR700550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 503610-43-3
Molekylformel C22 H26 F N3 O5 S
Molekylvikt (g/mol) 463.52
Synonym Methanesulfonamide, N-[4-(4-fluorophenyl)-6-(1-methylethyl)-5-[(1E)-2-[(2S,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethenyl]-2-pyrimidinyl]-N-methyl-, Rosuvastatin lactone, Rosuvastatin-5S-lactone, N-[4-(4-Fluorophenyl)-6-(1-methylethyl)-5-[(1E)-2-[(2S,4R)-tetrahydro-4-hydroxy-6-oxo-2H-pyran-2-yl]ethenyl]-2-pyrimidinyl]-N-methylmethanesulfonamide, N-[4-(4-Fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxotetrahydro-2H-pyran-2-yl]ethenyl]-6-(1-methylethyl)pyrimidin-2-yl]-N-methylmethanesulfonamide
IUPAC-namn N-[4-(4-fluorophenyl)-5-[(E)-2-[(2S,4R)-4-hydroxy-6-oxooxan-2-yl]ethenyl]-6-propan-2-ylpyrimidin-2-yl]-N-methylmethanesulfonamide
LEDER CC(C)c1nc(nc(c1\C=C\[C@@H]2C[C@@H](O)CC(=O)O2)c3ccc(F)cc3)N(C)S(=O)(=O)C
Specifikationer
Kemiskt namn eller material Rosuvastatin Lactone
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C22H26FN3O5S/c1-13(2)20-18(10-9-17-11-16(27)12-19(28)31-17)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)32(4,29)30/h5-10,13,16-17,27H,11-12H2,1-4H3/b10-9+/t16-,17-/m1/s1
Formel vikt 463.1577
Procent renhet >95%