Läs mer
Rosuvastatin Sodium Salt, TRC
Produktkod. 30410272
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30410272 500 mg
1 options
Produktkod. 30410272 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR225340500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Rosuvastatin Sodium
CAS 147098-18-8
Molekylformel C22 H27 F N3 O6 S . Na
Kvantitet 500 mg
Synonym 6-Heptenoic acid','7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-','sodium salt (1:1)','(3R,5S,6E)-, 6-Heptenoic acid','7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-','monosodium salt','(3R,5S,6E)- (9CI), 6-Heptenoic acid','7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-3,5-dihydroxy-','monosodium salt','[S-[R*,S*-(E)]]-, (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[(methanesulfonyl)methylamino]pyrimidin-5-yl]-3,5-dihydroxyhept-6-enoic acid sodium salt, (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl]-3,5-dihydroxy-6-heptenoic acid sodium salt, (3R,5S,6E)-7-[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl]-3,5-dihydroxy-6-heptenoic acid sodium salt, Rosuvastatin Sodium, Rosuvastatin sodium salt
InChI formel InChI=1S/C22H28FN3O6S.Na/c1-13(2)20-18(10-9-16(27)11-17(28)12-19(29)30)21(14-5-7-15(23)8-6-14)25-22(24-20)26(3)33(4,31)32;/h5-10,13,16-17,27-28H,11-12H2,1-4H3,(H,29,30);/q;+1/p-1/b10-9+;/t16-,17-;/m1./s1
LEDER [Na+].CC(C)c1nc(nc(c1\C=C\[C@@H](O)C[C@@H](O)CC(=O)[O-])c2ccc(F)cc2)N(C)S(=O)(=O)C
IUPAC-namn sodium;(E,3R,5S)-7-[4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]-3,5-dihydroxyhept-6-enoate
Molekylvikt (g/mol) 503.52
Formel vikt 503.15