Läs mer
Rosuvastatin Triphenylphosphonium Bromide, TRC
Produktkod. 30412525
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30412525 1 g
1 options
Produktkod. 30412525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR7005701G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Rosuvastatin Triphenylphosphonium Bromide
CAS 885477-83-8
Molekylformel C34 H34 F N3 O2 P S . Br
Kvantitet 1 g
Synonym Phosphonium','[[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]methyl]triphenyl-','bromide (1:1), Phosphonium','[[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]methyl]triphenyl-','bromide (9CI), [[4-(4-Fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]methyl]triphenylphosphonium bromide, ((4-(4-Fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)methyl)triphenylphosphonium bromide, Rosuvastatin triphenylphosphonium bromide
InChI formel InChI=1S/C34H34FN3O2PS.BrH/c1-25(2)32-31(33(26-20-22-27(35)23-21-26)37-34(36-32)38(3)42(4,39)40)24-41(28-14-8-5-9-15-28,29-16-10-6-11-17-29)30-18-12-7-13-19-30;/h5-23,25H,24H2,1-4H3;1H/q+1;/p-1
LEDER [Br-].CC(C)c1nc(nc(c1C[P+](c2ccccc2)(c3ccccc3)c4ccccc4)c5ccc(F)cc5)N(C)S(=O)(=O)C
IUPAC-namn [4-(4-fluorophenyl)-2-[methyl(methylsulfonyl)amino]-6-propan-2-ylpyrimidin-5-yl]methyl-triphenylphosphanium;bromide
Molekylvikt (g/mol) 678.59
Formel vikt 677.1277