Läs mer
Fisherbrand™ Rotary Vane Vacuum Pump
Produktkod. 12911141 Handla allt Fisherbrand Traceable Produkter
Change view
Flödeshastighet:
120 till 143 l/min
30 till 36 l/min
70 till 92 l/min
97 till 110 l/min
Förpackningsstorlek:
Varje
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Flödeshastighet Förpackningsstorlek
12911141 70 till 92 l/min Varje
12931141 120 till 143 l/min Varje
12901141 30 till 36 l/min Varje
12921141 97 till 110 l/min Varje
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 12911141
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Designed for laboratory applications requiring deep vacuum. Fisherbrand™ Rotary Vane Vacuum Pump is a two-stage direct driven rotary vane pumps that provide a high water vapor tolerance.
  • Two-stage direct driven rotary vane pump
  • Designed for wide range of physical and chemical laboratory applications and continuous running
  • Low weight, small footprint and high pumping speed, increased chemical resistance are advantageous features in research and routine processes
  • Rugged reliable design, combined with low noise level and ease of use
  • Innovative gas ballast valve prevents moisture condensing and high tolerance of water and solvent vapors
  • Intake line is vacuum sealed by and anti suckback valve to protect your application from undesired venting and oil back flow after pump is turned off
  • Vertical oil sight glass allows easy monitoring of oil level and condition
TRUSTED_SUSTAINABILITY
Specifikationer
Max. Vakuum 2 x 10-3 mbar
Elektriska krav 115/230 V 50/60 Hz motor with Schuko UK and US power leads
Mått (L x B x H) 41.5 x 15.0 x 23.5 cm (16.3 x 5.9 x 9.25 in.)
Wattal 90 w
Vikt (engelska) 38.5 lb.
Vikt (metrisk) 17.5 kg
Flödeshastighet 70 till 92 l/min
Frekvens 50/60 Hz
Pumptyp Rotary Vane Vacuum Pump
Luftförskjutning 77 L/min.
Användningsområden Backing Pump for UHV Applications
Maximalt tryck 2 x 10-3 mbar
För användning med (utrustning) Labovac 10
Spänning 230 V