Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
670.00 SEK - 1680.00 SEK
Specifications
Type | Adapter |
---|---|
For Use With (Equipment) | Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge |
Product Code | Brand | Tube Type | Capacity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Capacity | Price | Quantity & Availability | ||||
10392352
|
Hettich™
1437 |
Round Bottom | 1 x 100 mL Tube |
720.00 SEK
Each |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10318343
|
Hettich™
1438 |
Conical | 7 x 1.6 to 7 mL Tubes |
1030.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10659792
|
Hettich™
1431 |
Round Bottom | 7 x 15 mL |
740.00 SEK
Each |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10033942
|
Hettich™
1737 |
Conical Skirted | 50 mL Tubes |
1330.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12053874
|
Hettich™
1433 |
Round Bottom | 14 x 3 mL |
1680.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10272293
|
Hettich™
1434 |
Round Bottom | 12 x 6 to 7 mL Tubes |
1540.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10344443
|
Hettich™
1439 |
Round Bottom | 4 x 9 to 10 mL Tubes |
1030.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10246903
|
Hettich™
1441 |
Conical, Blood/Urine | 7 x 4 to 10 mL Tubes |
1050.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10347322
|
Hettich™
1443 |
Conical | 50 mL |
670.00 SEK
Each |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Rotor adapter for Hettich Carriers
- For use with Universal 320/320 R Centrifuge
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Adapter | |
Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge |