Learn More
Hettich™ Rotor Adapter for Carriers no. 1366 and 1427
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1327
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12053864
  • 1040.00 SEK / Each
Estimated Shipment: 08-09-2021
Description
Description
  • Rotor adapter for Hettich carriers
  • For use with Rotina, Rotofix and Universal Centrifuges
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
  • ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
  • ROTINA Centrifuges are ideally equipped to deliver separation technology to satisfy the highest standards
Specifications
Specifications
12 x 3 mL Tubes
Hettich Rotors with Carriers no. 1366 and 1427
Rotor Adapter
12