Learn More
SVL™ Rotulex™ Spherical Joints
Offers a grease free spherical joint
Brand:  SVL™ RM41
 View more versions of this product
Product Code. 15507960
  • 20070.00 SEK / Pack of 30
Only null left
Estimated Shipment: 23-10-2025
Description
  • All glass and PTFE pathway is highly resistant to chemical attack
  • Can be used at temperatures up to 200°C
  • System comprises male ball joint with PTFE covered O-ring which fits into female cup joint
  • Joint can be retained together with spring loaded metal clip
  • Component parts RM, RF, RO and RC should be ordered separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Male Joint, PTFE Covered O-ring, Spring-loaded Metal clip
Glass, PTFE
Spherical joint
41
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SVL™ Rotulex™ Spherical Joints > Male; Bore size: 25mm; Joint size: 41