Läs mer
RP 48497 (Eszopiclone Impurity C), TRC
Produktkod. 30135278
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135278 100 mg
30006358 25 mg
30012553 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135278 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR700900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 148891-53-6
Molekylformel C11 H7 Cl N4 O
Molekylvikt (g/mol) 246.65
Synonym 6-(5-Chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one, Zopiclone Imp. C (EP)
IUPAC-namn 6-(5-chloropyridin-2-yl)-7H-pyrrolo[3,4-b]pyrazin-5-one
LEDER Clc1ccc(nc1)N2Cc3nccnc3C2=O
Specifikationer
Kemiskt namn eller material 6-(5-Chloropyridin-2-yl)-6,7-dihydro-5H-pyrrolo[3,4-b]pyrazin-5-one
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C11H7ClN4O/c12-7-1-2-9(15-5-7)16-6-8-10(11(16)17)14-4-3-13-8/h1-5H,6H2
Formel vikt 246.0308
Procent renhet >95%