Läs mer
Tocris Bioscience™ RP 6306
GREENER_CHOICE
Produktkod. 30000445
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30000445 10 mg
30000428 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30000445 Leverantör Tocris Bioscience™ Leverantörsnummer 8031/10
Potent and selective PKMYT1 inhibitor
RP 6306 is a potent and selective PKMYT1 inhibitor (IC50 = 3.1 nM). In vivo, it inhibits CCNE1-amplified tumor cell growth in mouse xenograft models. RP 6306 is orally bioavailable.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-2-Amino-1-(3-hydroxy-2,6-dimethylphenyl)-5,6-dimethylpyrrolo[2,3-b]pyridine-3-carboxamide
CAS 2719793-90-3
Kvantitet 10 mg
Mål Cyclin-dependent Kinase Inhibitors
Molekylformel C18H20N4O2
Renhet 98%