Läs mer
RPA 202248, TRC
Produktkod. 30136032
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136032 25 mg
17960465 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136032 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR70091525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 143701-75-1
Molekylformel C15 H12 F3 N O4 S
Molekylvikt (g/mol) 359.32
Synonym Benzenepropanenitrile, α-(cyclopropylcarbonyl)-2-(methylsulfonyl)-β-oxo-4-(trifluoromethyl)-, α-(Cyclopropylcarbonyl)-2-(methylsulfonyl)-β-oxo-4-(trifluoromethyl)benzenepropanenitrile, RPA 202248
IUPAC-namn 2-(cyclopropanecarbonyl)-3-[2-methylsulfonyl-4-(trifluoromethyl)phenyl]-3-oxopropanenitrile
LEDER CS(=O)(=O)c1cc(ccc1C(=O)C(C#N)C(=O)C2CC2)C(F)(F)F
Specifikationer
Kemiskt namn eller material Isoxaflutole-diketonitrile
Kvantitet 25 mg
InChI formel InChI=1S/C15H12F3NO4S/c1-24(22,23)12-6-9(15(16,17)18)4-5-10(12)14(21)11(7-19)13(20)8-2-3-8/h4-6,8,11H,2-3H2,1H3
Formel vikt 359.0439