Learn More
RPA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564256
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MSKSGFGSYG SISAADGASG GSDQLCERDA TPAIKTQRPK VRIQDVVPCN VNQLLSSTVF DPVFKVRGII VSQV Highest antigen sequence identity to the following orthologs: Mouse - 38%, Rat - 41%.
RPA4 Polyclonal antibody specifically detects RPA4 in Human samples. It is validated for Immunohistochemistry.Specifications
RPA4 | |
Polyclonal | |
Unconjugated | |
RPA4 | |
HSU24186; MGC120333; MGC120334; Replication factor A protein 4; replication protein A 30 kDa subunit; replication protein A complex 34 kd subunit homolog Rpa4; replication protein A4; replication protein A4, 30kDa; replication protein A4, 34kDa; RF-A protein 4; RP-A p30; RPA4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29935 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13156 | |
RPA4 | |
Recombinant protein corresponding to Human RPA4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only