Learn More
uvex RPL Hypervision CB Clear AF
Product Code. 31751173 Shop All Uvex Products
Change view
Click to view available options
Lens Material:
Polycarbonate
Unit Size:
Each
Product Code. Lens Material Unit Size
31751173 Polycarbonate Each
1 options
Product Code. 31751173 Supplier uvex Supplier No. 9321955
  • ONLINE EXCLUSIVE PRICE
    127.39 SEK
    120.93 SEK / Paket med 1
    Save 6.46 SEK
    5% Av
Beräknad leverans: 16-11-2026
For reliable visual clarity and protection.
  • RPL Hypervision CB eyewear featuring a clear AF lens, designed to deliver dependable visual clarity and consistent performance, identified as Hypervision CB clear AF.
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant, Impact-resistant, Anti-fog
Lens Tint None
Product Type Replacement Lens
Disposable Reusable
Lens Material Polycarbonate
Autoclavable Not Autoclavable