missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL26L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RPL26L1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836134
|
Proteintech
16487-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826134
|
Proteintech
16487-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein that shares high sequence similarity with ribosomal protein L26. Alternative splicing results in multiple transcript variants encoding the same protein.Specifications
RPL26L1 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q9UNX3 | |
307636, 51121, 668405 | |
RPL26L1 Fusion Protein Ag9609 | |
Primary | |
-20°C | |
RPL26L1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ribosomal protein L26 like 1, RPL26L1, RPL26P1 | |
RPL26L1, Rpl26-ps3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |