missing translation for 'onlineSavingsMsg'
Learn More

RPL28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 16649-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16826344

  • 3313.85 SEK / 150µL
Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L28E family of ribosomal proteins. It is located in the cytoplasm. Variable expression of this gene in colorectal cancers compared to adjacent normal tissues has been observed, although no correlation between the level of expression and the severity of the disease has been found. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
Specifications

Specifications

RPL28
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17702, P41105, P46779
60S ribosomal protein L28, ribosomal protein L28, RPL28
Rabbit
IgG
150 μL
-20°C
Primary
19943, 6158, 64638
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
RPL28
Liquid
RPL28
RPL28 Fusion Protein Ag10036
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers