Learn More
RPL39L Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24940-1-AP-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16823465
  • 1236.20 SEK / 20µL
Estimated Shipment: 08-12-2021
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.
Specifications
Specifications
RPL39L
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96EH5
60S ribosomal protein L39 2, 60S ribosomal protein L39 like, ribosomal protein L39 like, RPL39L, RPL39L1
Rabbit
IgG
20 μL
-20°C
Primary
116832
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
RPL39L
Liquid
RPL39L
RPL39L Fusion Protein Ag20923
Antigen Affinity Chromatography