missing translation for 'onlineSavingsMsg'
Learn More

RPL9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14117-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16832614

  • 1656.73 SEK / 20µL
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 60S subunit. The protein belongs to the L6P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

RPL9
Polyclonal
Unconjugated
RPL9
60S ribosomal protein L9, ribosomal protein L9, RPL9, RPL9P7, RPL9P8, RPL9P9
Rabbit
Antigen Affinity Chromatography
RUO
20005, 29257, 6133
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17077, P32969, P51410
RPL9
RPL9 Fusion Protein Ag5266
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers