Learn More
RPS10 Mouse anti-Human, Unconjugated, Clone: 1G1E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67609-1-IG-20UL
This item is not returnable.
View return policy
Description
The RPS10 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPS10. This antibody recognizes human, mouse, rat antigen. The RPS10 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
RPS10 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, ELISA | |
1G1E1 | |
Unconjugated | |
20 μL | |
6204 | |
IgG2b |