missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ RPS17 Recombinant Protein
Brand: Abnova™ H00006218-P01.2ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Antibody Production, ELISA, Protein Array, Western Blot | |
40.59 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
2μg | |
-80°C | |
Recombinant |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
Human RPS17 Full-length ORF Recombinant Protein with GST-tag at N-terminal | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE stained with Coomassie Blue | |
Wheat Germ (in vitro) | |
Human | |
Solution |