missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS28 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14796-1-AP-20UL
Additional Details : Weight : 0.00010kg
Descrizione
TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.Specifica
RPS28 | |
Polyclonal | |
Unconjugated | |
RPS28 | |
40S ribosomal protein S28, ribosomal protein S28, RPS28 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54127, 6234, 691531 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62857, P62858, P62859 | |
Rps28 | |
RPS28 Fusion Protein Ag6505 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |